ENA117737852 Enamine

N-benzyl-5-(hydroxyimino)-2-oxo-1,2,5,6,7,8-hexahydroquinoline-3-carboxamide

  • Cas Number : 940242-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27784

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27784
Availability details for
Quantity
  • Availability
  • Add to Cart