ENA117737855 Enamine

1-(2,4,5-trimethylthieno[2,3-d]pyrimidin-6-yl)ethanone

  • MDL number MFCD09396739
  • Cas Number : 941083-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27801

Safety Information

msds https://www.enaminestore.com/catalog/EN300-27801
Availability details for
Quantity
  • Availability
  • Add to Cart