ENA117737857 Enamine

3-(morpholin-4-yl)-3-oxo-2-[5-(trifluoromethyl)pyridin-2-yl]propanenitrile

  • MDL number MFCD09396713
  • Cas Number : 941133-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-27807

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27807
Availability details for
Quantity
  • Availability
  • Add to Cart