ENA117737861 Enamine

1-[(1-methyl-1H-pyrrol-3-yl)sulfonyl]piperidine-4-carboxylic acid

  • MDL number MFCD09389740
  • Cas Number : 940271-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27820

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27820
Availability details for
Quantity
  • Availability
  • Add to Cart