ENA117737868 Enamine

7-fluoro-2,3,4,5-tetrahydro-1-benzoxepin-5-one

  • MDL number MFCD08446939
  • Cas Number : 774-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 46-47
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27863

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27863
Availability details for
Quantity
  • Availability
  • Add to Cart