ENA117737869 Enamine

2-methylpyrimidine-4-carbaldehyde oxime

  • MDL number MFCD09702180
  • Cas Number : 28840-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-27873
Availability details for
Quantity
  • Availability
  • Add to Cart