ENA117737872 Enamine

1,3-dimethyl-5-[(methylamino)methyl]-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD09392375
  • Cas Number : 941209-04-7
  • Molecular Weight : 183.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-28235

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28235
Availability details for
Quantity
  • Availability
  • Add to Cart