ENA117737875 Enamine

5-(trifluoromethyl)-1-[5-(trifluoromethyl)pyridin-2-yl]-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD09403616
  • Cas Number : 1013794-64-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-28263

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28263
Availability details for
Quantity
  • Availability
  • Add to Cart