ENA117737876 Enamine

1-(6-methoxypyridazin-3-yl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD09403614
  • Cas Number : 1013794-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28264

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28264
Availability details for
Quantity
  • Availability
  • Add to Cart