ENA117737892 Enamine

2-(furan-2-yl)-4-methyl-6-(methylsulfanyl)pyrimidine-5-carboxylic acid

  • MDL number MFCD09403768
  • Cas Number : 941096-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28440

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28440
Availability details for
Quantity
  • Availability
  • Add to Cart