ENA117737893 Enamine

2-tert-butyl-4-mercapto-6-methylpyrimidine-5-carboxylic acid

  • MDL number MFCD09403623
  • Cas Number : 941117-68-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28443

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28443
Availability details for
Quantity
  • Availability
  • Add to Cart