ENA117737906 Enamine

phenyl 2-acetylphenylcarbamate

  • MDL number MFCD09413158
  • Cas Number : 941378-46-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28567

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28567
Availability details for
Quantity
  • Availability
  • Add to Cart