ENA117737908 Enamine

2-(1H-1,2,4-triazol-1-yl)pyridine-3-carboxylic acid

  • MDL number MFCD09413277
  • Cas Number : 941400-53-9
  • Molecular Weight : 190.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(1H-1,2,4-triazol-1-yl)nicotinic acid
Inchi Code 1S/C8H6N4O2/c13-8(14)6-2-1-3-10-7(6)12-5-9-4-11-12/h1-5H,(H,13,14)
Inchi Key JVRBJJCNDSCZJV-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-28571

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-28571
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart