ENA117737913 Enamine

2-azepan-1-yl-3-methylbutan-1-amine

  • MDL number MFCD09259718
  • Cas Number : 929343-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28596

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28596
Availability details for
Quantity
  • Availability
  • Add to Cart