ENA117737914 Enamine

3-methyl-2-(4-methylpiperazin-1-yl)butan-1-amine

  • MDL number MFCD09259724
  • Cas Number : 929343-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28597

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28597
Availability details for
Quantity
  • Availability
  • Add to Cart