ENA117737919 Enamine

2-chloro-4-(4-fluorophenyl)-1,3-thiazole

  • MDL number MFCD08446728
  • Cas Number : 42445-38-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-28614

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28614
Availability details for
Quantity
  • Availability
  • Add to Cart