ENA117737921 Enamine

1-[4-(1H-1,2,4-triazol-1-yl)phenyl]ethanone oxime

  • MDL number MFCD09413276
  • Cas Number : 941400-50-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28760

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28760
Availability details for
Quantity
  • Availability
  • Add to Cart