ENA117737959 Enamine

N-[(2-chloro-6-fluorophenyl)methyl]-N-methylsulfamoyl chloride

  • MDL number MFCD08444959
  • Cas Number : 923688-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-28270

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28270
Availability details for
Quantity
  • Availability
  • Add to Cart