ENA117737963 Enamine

3-(5-bromo-1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid

  • MDL number MFCD00574198
  • Cas Number : 299964-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-22734

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-22734
Availability details for
Quantity
  • Availability
  • Add to Cart