ENA117737969 Enamine

1-(3-fluorophenyl)cyclobutanecarboxylic acid

  • MDL number MFCD08444832
  • Cas Number : 179411-84-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 67...69
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28290

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28290
Availability details for
Quantity
  • Availability
  • Add to Cart