ENA117737978 Enamine

N-allyl-N'-(4-nitrophenyl)urea

  • MDL number MFCD00024607
  • Cas Number : 194788-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28434

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28434
Availability details for
Quantity
  • Availability
  • Add to Cart