ENA117737979 Enamine

N-(4-nitrophenyl)-N'-(tetrahydrofuran-2-ylmethyl)urea

  • MDL number MFCD07044883
  • Cas Number : 903708-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27787

Safety Information

msds https://www.enaminestore.com/catalog/EN300-27787
Availability details for
Quantity
  • Availability
  • Add to Cart