ENA117737982 Enamine

2-(oxolan-2-yl)ethan-1-amine

  • MDL number MFCD07329647
  • Cas Number : 98277-97-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27797

Safety Information

signalword Warning
pcodes P264,P270,P280,P301+P312,P305+P351+P338,P330,P337+P313,P501
MSDS https://www.enaminestore.com/catalog/EN300-27797
pictograms GHS07
hstatements H302,H319
Availability details for
Quantity
  • Availability
  • Add to Cart