ENA117737986 Enamine

1-(2-fluorophenyl)-2,5-dimethyl-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD08444945
  • Cas Number : 923781-31-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28445

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28445
Availability details for
Quantity
  • Availability
  • Add to Cart