ENA117737993 Enamine

N-(piperidin-4-yl)cyclopropanecarboxamide

  • MDL number MFCD05254808
  • Cas Number : 836685-68-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-28527

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28527
Availability details for
Quantity
  • Availability
  • Add to Cart