ENA117737995 Enamine

3-chloro-N-cyclohexylpropanamide

  • MDL number MFCD02285967
  • Cas Number : 61872-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25813

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25813
Availability details for
Quantity
  • Availability
  • Add to Cart