ENA117737998 Enamine

3-(1H-1,2,4-triazol-1-yl)propanenitrile

  • MDL number MFCD03727265
  • Cas Number : 76686-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28535

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28535
Availability details for
Quantity
  • Availability
  • Add to Cart