ENA117738006 Enamine

8-fluoro-5-nitroquinoline

  • MDL number MFCD09048716
  • Cas Number : 94832-39-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-27811

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27811
Availability details for
Quantity
  • Availability
  • Add to Cart