ENA117738015 Enamine

4-carbamothioylbenzoic acid

  • MDL number MFCD09044819
  • Cas Number : 4989-36-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28564

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28564
Availability details for
Quantity
  • Availability
  • Add to Cart