ENA117738023 Enamine

6-[(hydroxyimino)methyl]-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • Cas Number : 81450-61-5
  • Molecular Weight : 163.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28575

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28575
Availability details for
Quantity
  • Availability
  • Add to Cart