ENA117738031 Enamine

5-{[3,5-bis(trifluoromethyl)phenyl]methyl}-1,3-thiazol-2-amine

  • MDL number MFCD09051004
  • Cas Number : 937631-44-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-28588

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28588
Availability details for
Quantity
  • Availability
  • Add to Cart