ENA117738032 Enamine

4-[(2-amino-1,3-thiazol-5-yl)methyl]benzonitrile

  • MDL number MFCD08442697
  • Cas Number : 926253-88-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-28591

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28591
Availability details for
Quantity
  • Availability
  • Add to Cart