ENA117738033 Enamine

(6-bromo-1H-indol-2-yl)methanol

  • MDL number MFCD08445239
  • Cas Number : 923197-75-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112-114
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27831

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27831
Availability details for
Quantity
  • Availability
  • Add to Cart