ENA117738051 Enamine

3-(2-imino-4-methyl-2,3-dihydro-1,3-thiazol-5-yl)-4-methyl-4,5-dihydro-1H-1,2,4-triazole-5-thione

  • MDL number MFCD23145023
  • Cas Number : 587007-13-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-28744

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28744
Availability details for
Quantity
  • Availability
  • Add to Cart