ENA117738053 Enamine

2-methyl-5-(methylthio)benzoic acid

  • MDL number MFCD09702196
  • Cas Number : 26246-30-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128...130
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28747

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28747
Availability details for
Quantity
  • Availability
  • Add to Cart