ENA117738057 Enamine

1-[4-(1H-imidazol-1-yl)phenyl]ethanone oxime

  • MDL number MFCD00127064
  • Cas Number : 146139-83-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28762

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28762
Availability details for
Quantity
  • Availability
  • Add to Cart