ENA117738070 Enamine

6-chloro-8-nitro-3,4-dihydro-2H-1-benzopyran-3-carboxylic acid

  • MDL number MFCD08442672
  • Cas Number : 926239-75-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27865

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27865
Availability details for
Quantity
  • Availability
  • Add to Cart