ENA117738077 Enamine

2-(trifluoromethyl)cyclohexanamine

  • MDL number MFCD09040726
  • Cas Number : 58665-69-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27688

Safety Information

msds https://www.enaminestore.com/catalog/EN300-27688
Availability details for
Quantity
  • Availability
  • Add to Cart