ENA117738085 Enamine

1-(3-chlorophenyl)cyclobutane-1-carboxylic acid

  • MDL number MFCD03408895
  • Cas Number : 151157-55-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28233

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28233
Availability details for
Quantity
  • Availability
  • Add to Cart