ENA117738096 Enamine

3-[(2,4-dioxo-1,3-thiazolidin-3-yl)methyl]benzoic acid

  • MDL number MFCD03131888
  • Cas Number : 488823-77-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-27715

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27715
Availability details for
Quantity
  • Availability
  • Add to Cart