ENA117738103 Enamine

1-chloro-3-phenylpropan-2-ol

  • MDL number MFCD00093259
  • Cas Number : 5396-65-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27722

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27722
Availability details for
Quantity
  • Availability
  • Add to Cart