ENA118036413 Enamine

2-{3-oxo-2H,3H-[1,2,4]triazolo[4,3-a]pyridin-2-yl}acetic acid

  • MDL number MFCD09396709
  • Cas Number : 941055-65-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27476

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27476
Availability details for
Quantity
  • Availability
  • Add to Cart