ENA118036415 Enamine

6-chloro-1-methyl-1H,2H,3H,4H-pyrrolo[1,2-a]pyrazine

  • Molecular Weight : 170.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-chloro-1-methyl-1,2,3,4-tetrahydropyrrolo[1,2-a]pyrazine
Inchi Code 1S/C8H11ClN2/c1-6-7-2-3-8(9)11(7)5-4-10-6/h2-3,6,10H,4-5H2,1H3
Inchi Key XXVWOUMHIVZHED-UHFFFAOYSA-N

Description

Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-27651
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart