ENA118036419 Enamine

1-(1-azabicyclo[2.2.2]oct-3-yl)-2,5-dimethyl-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD09802056
  • Cas Number : 1000930-76-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28524

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28524
Availability details for
Quantity
  • Availability
  • Add to Cart