ENA118036423 Enamine

2-[(1-chloro-2,2,2-trifluoroethyl)sulfanyl]aniline

  • MDL number MFCD09802064
  • Cas Number : 949259-77-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28771

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28771
Availability details for
Quantity
  • Availability
  • Add to Cart