ENA118036428 Enamine

1-{5-methyl-1-[5-(trifluoromethyl)pyridin-2-yl]-1H-pyrazol-4-yl}ethanone oxime

  • MDL number MFCD09802070
  • Cas Number : 1019075-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28801

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28801
Availability details for
Quantity
  • Availability
  • Add to Cart