ENA118036433 Enamine

2-[(4-chlorophenoxy)methyl]-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD09802074
  • Cas Number : 949259-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28810

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28810
Availability details for
Quantity
  • Availability
  • Add to Cart