ENA118036434 Enamine

methyl 3-(cyanomethyl)furan-2-carboxylate

  • MDL number MFCD09802075
  • Cas Number : 68846-20-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28812

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28812
Availability details for
Quantity
  • Availability
  • Add to Cart