ENA118036440 Enamine

2-methyl-7,8,9,10-tetrahydro-6H-pyrido[3',2':4,5]imidazo[1,2-a]azepine-4-carboxylic acid

  • MDL number MFCD09802081
  • Cas Number : 949654-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 274...276
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28824

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28824
Availability details for
Quantity
  • Availability
  • Add to Cart