ENA118036442 Enamine

3-bromo-5-(trichloroacetyl)-1H-pyrrole-2-sulfonyl chloride

  • MDL number MFCD09802086
  • Cas Number : 949204-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28881

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28881
Availability details for
Quantity
  • Availability
  • Add to Cart